Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Recombinant Mouse Histone Deacetylase 8/HDAC8 His Protein
Shop All Novus Biologicals Products

Click to view available options
Quantity:
10 μg
50 μg
Description
A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 1-377 of Mouse Histone Deacetylase 8/HDAC8 The Recombinant Mouse Histone Deacetylase 8/HDAC8 Protein is derived from E. coli. The Recombinant Mouse Histone Deacetylase 8/HDAC8 Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
Concentration | 0.25mg/mL |
For Use With (Application) | SDS-PAGE |
Formulation | Liquid. PBS (pH 7.4) containing 10% glycerol |
Gene ID (Entrez) | 55869 |
Molecular Weight (g/mol) | 42.5 kDa |
Name | Recombinant Human Histone Deacetylase 8/HDAC8 Protein |
Purification Method | >90%, by SDS-PAGE |
Quantity | 50 μg |
Source | Baculovirus |
Storage Requirements | Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles. |
Show More |
For Research Use Only.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction