Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Mouse IL-12 p70/IL-12A Protein
SDP

Catalog No. NBP2762671M Shop All R&D Systems Products
Click to view available options
Quantity:
1 mg
2 μg

Highly purified and high bioactivity. Generating reliable and reproducible results.

An un-tagged bioactive recombinant protein corresponding to Mouse IL-12 p70/IL-12A

The Recombinant Mouse IL-12 p70/IL-12A Protein has been validated for the following applications: SDS-Page, Bioactivity.

Specifications

Concentration Lyoph
For Use With (Application) Bioactivity, SDS-PAGE
Gene ID (Entrez) 3592
Molecular Weight (g/mol) 57.6 kDa
Purification Method >95%, by SDS-PAGE
Quantity 1 mg
Source HEK293
Storage Requirements Store at -20 to -80°C. Avoid freeze-thaw cycles.
Gene Symbol IL12A
Specific Reactivity IL-12 p70/IL-12A is a heterodimer, p35: 21.8 KDa, p40: 35.8 kDa total: 57.6 kDa (p35: 194 aa, p40: 313 aa total: 507 aa aa)
Reconstitution Sterile water at 0.1 mg/mL
Buffer Lyophilized from a sterile (0.2 micron) filtered aqueous solution containing 10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.