Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Recombinant Mouse IL-17/IL-17A Protein
Shop All Novus Biologicals Products

Click to view available options
Quantity:
1 mg
Description
Production of IL-6 by mouse 3T3 cells, >2.0 x 10^5 units/mg (ED50 acceptance criteria are <5 ng/mL).
Specifications
Specifications
For Use With (Application) | Bioactivity, SDS-PAGE |
Formulation | Lyophilized from a 0.2 micron filtered solution, 0.1% Trifluoroacetic Acid (TFA) |
Gene ID (Entrez) | 3605 |
Quantity | 1 mg |
Storage Requirements | Store at -20 to -70C as supplied. After reconstitution, store at 2 to 8C for 1 month and at -20 to -70C for long term storage. Avoid repeated freeze-thaw cycles. |
Endotoxin Concentration | Acceptance criteria for endotoxin levels are <1 EUs/μg protein (determined by Kinetic LAL Method). |
Gene Alias | CTLA8cytotoxic T-lymphocyte-associated serine esterase 8, Cytotoxic T-lymphocyte-associated antigen 8, IL-17Acytotoxic T-lymphocyte-associated protein 8, IL-17CTLA-8, IL17interleukin-17A, interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8), interleukin 17A |
Gene Symbol | IL17A |
Reconstitution | Sterile water at 0.1 mg/mL |
Research Category | Apoptosis, Biologically Active Proteins, Cytokine Research, Immunology, Innate Immunity |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction