Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Recombinant Mouse M-CSF Protein
Shop All Novus Biologicals Products

Click to view available options
Quantity:
1 mg
500 μg
Description
NFS-60 cell proliferation, >1.0 x 10^5 units/mg (ED50 acceptance criteria are <10 ng/mL).
Specifications
Specifications
For Use With (Application) | Bioactivity, SDS-PAGE |
Formulation | Lyophilized from a 0.2 micron filtered solution, 10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5 |
Gene ID (Entrez) | 1435 |
Quantity | 500 μg |
Storage Requirements | Store at -20 to -70C as supplied. After reconstitution, store at 2 to 8C for 1 month and at -20 to -70C for long term storage. Avoid repeated freeze-thaw cycles. |
Endotoxin Concentration | Acceptance criteria for endotoxin levels are <1 EUs/μg protein (determined by Kinetic LAL Method). |
Gene Alias | colony stimulating factor 1 (macrophage), CSF-1, Lanimostim, macrophage colony stimulating factor, macrophage colony-stimulating factor 1, M-CSF, MCSFlanimostim, MGC31930 |
Gene Symbol | CSF1 |
Reconstitution | Sterile water at 0.1 mg/mL |
Research Category | Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research, Stem Cell Markers |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction