Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Recombinant Mouse PKM2 Protein
Shop All Novus Biologicals Products

Click to view available options
Quantity:
100 μg
20 μg
Description
One unit will convert 1.0 pmole of phospho(enol)pyruvate to pyruvate per minute at pH 7.5 at 37C
Specifications
Specifications
Accession Number | NP_035229, NP_035229 |
Concentration | 0.5 mg/ml |
Formulation | 20 mM Tris-HCl buffer (pH 8.5), 0.2 M NaCl, 1 mM DTT, 30% glycerol |
Gene ID (Entrez) | 5315 |
Molecular Weight (g/mol) | Theroretical: 60.2 kDa |
Quantity | 100 μg |
Storage Requirements | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Gene Symbol | PKM |
Purity or Quality Grade | >85%, by SDS-PAGE |
Protein | PKM2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction