Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Mouse RalA/RalB Protein
SDP

Catalog No. p-7145976 Shop All R&D Systems Products
Click to view available options
Quantity:
0.5 mg
100 μg

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids1-203 of MouseRalA/RalB The Recombinant Mouse RalA/RalB Protein is derived from E. coli. The Recombinant Mouse RalA/RalB Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 0.5 mg/ml
For Use With (Application) SDS-PAGE
Formulation PBS (pH 7.4) containing 10% glycerol.
Gene ID (Entrez) 5898
Molecular Weight (g/mol) 25.7 kDa
Quantity 100 μg
Source E. coli
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Gene Symbol RALA
Cross Reactivity RalA/RalB
Species Mouse
Purity or Quality Grade >90%, by SDS-PAGE
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.