Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Rat IL-2 Animal-Free Protein
SDP

Catalog No. NBP2761871M Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
2 μg
20 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP2761871M 1 mg
NBP2761872U 2 μg
NB27618720U 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NBP2761871M Supplier Novus Biologicals™ Supplier No. NBP2761871MG

Highly purified and high bioactivity. Generating reliable and reproducible results.

An un-tagged bioactive recombinant protein corresponding to Rat IL-2

The Recombinant Rat IL-2 Animal-Free Protein has been validated for the following applications: SDS-Page, Bioactivity.

Specifications

Concentration Lyoph
For Use With (Application) Bioactivity, SDS-PAGE
Gene ID (Entrez) 3558
Molecular Weight (g/mol) 15.6 kDa
Purification Method >95%, by SDS-PAGE
Quantity 1 mg
Source E. coli
Storage Requirements Store at -20 to -80°C. Avoid freeze-thaw cycles.
Gene Symbol IL2
Specific Reactivity IL-2 is a monomer, 15.6 kDa (136 aa)
Reconstitution Sterile 10 mM HCl at 0.1 mg/mL
Buffer Lyophilized from a sterile (0.2 micron) filtered aqueous solution containing 0.1% Trifluoroacetic Acid (TFA)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.