Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Recombinant S. aureus Glutamyl endopeptidase Protein
Shop All R&D Systems Products

Click to view available options
Quantity:
1 mg
5 mg
Description
An un-tagged recombinant protein corresponding to S. aureus Glutamyl endopeptidase
The Recombinant S. aureus Glutamyl endopeptidase Protein has been validated for the following applications: SDS-Page.
The Recombinant S. aureus Glutamyl endopeptidase Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
Concentration | Lyoph |
For Use With (Application) | SDS-PAGE |
Molecular Weight (g/mol) | 28.9 kDa |
Purification Method | >95%, by SDS-PAGE |
Quantity | 5 mg |
Source | E. coli |
Storage Requirements | Store at -20 to -80°C. Avoid freeze-thaw cycles. |
Specific Reactivity | Glutamyl endopeptidase is a monomer, 28.9 kDa (267 aa) |
Reconstitution | Sterile water at 0.1 mg/mL |
Buffer | Lyophilized from a sterile (0.2 micron) filtered aqueous solution containing 10 mM sodium phosphate, pH 7.5 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction