Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Western Blot Protein Standards (12-90 kDa)
SDP

Catalog No. NB397961 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
250 Tests
50 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB397961 50 Tests
NB397962 250 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB397961 Supplier Novus Biologicals™ Supplier No. NBP31165050TESTS
Add to Cart
Add to Cart

Recombinant Protein

Specifications

Buffer 125 mM Tris-HCl, pH 6.8, 10 mM DTT, 2 mM EDTA, 2 % (W/V) SDS, 10 % (W/V) Glycerol, 0.01% (W/V) Bromophenol blue
Content And Storage Store at -20°C.
For Use With (Application) Western Blot
Quantity 50 Tests
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.