Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RECQ1 Antibody, Novus Biologicals™
SDP

Catalog No. NB100619 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100619 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100619 Supplier Novus Biologicals Supplier No. NB100619
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

RECQ1 Polyclonal specifically detects RECQ1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen RECQ1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunohistochemistry 1:2000-1:10000, Immunoprecipitation 2-10 μg/mg lysate, Immunohistochemistry-Paraffin 1:2000-1:10000
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. P46063
Gene Alias DNA helicase, RecQ-like type 1, DNA-dependent ATPase Q1, EC 3.6.1, EC 3.6.4.12, RecQ protein-like (DNA helicase Q1-like), RecQ protein-like 1, RECQ1, RecQ1ATP-dependent DNA helicase Q1, RecQL1
Gene Symbols RECQL
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 600 and the C-terminus (residue 649) of human RecQ protein-like (DNA helicase Q1-like) 1 using the numbering given in Swiss-Prot entry P46063 (GeneID 5965).
Molecular Weight of Antigen 73 kDa
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5965
Test Specificity Human. Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, Northern white-cheeked gibbon, Small-eared galago, and Thirteen-lined ground squirrel.
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.