Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Red Kap™ Women's Button-Front Lab Coats

Catalog No. 19166221 Shop All Red Kap Industries Products
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
Style:
Long
Regular
Short
This item is not returnable. View return policy
This item is not returnable. View return policy

Wrinkle-resistant with button closure lab coat features princess seams on the front and back for a flattering feminine fit.

  • 80% polyester/20% combed cotton blend
  • Pre-cure durable press finish
  • One-piece, lined, notched lapel collar
  • Left breast pocket, bar-tacked pencil stall, two lower pockets
  • Side vent openings; back has princess seams
  • White

Specifications

Product Type General Purpose Lab Coat
Size Small
Gender Women's
Material Polyester, Cotton
Color White
Closure Type Buttons
Disposable Reusable
Sterility Non-sterile
Style Short
No. of Pockets 3
Collar Type Notched Lapel
Finish Pre-cure Durable Press
Size (English) Chest Size 6 (33.5 to 34 in.), Size 8 (34.5 to 35 in.)
Size (English) Hip Size 6 (35 to 35.5 in.), Size 8 (36 to 36.5 in.)
Size (English) Waist Size 6 (25 to 25.5 in.), Size 8 (26 to 26.5 in.)
Size (Metric) Hip 88.9 to 90.17 (Size 6), 91.44 to 92.71 (Size 8)cm
Size (Metric) Waist Size 6 (63.5 to 64.77 cm), Size 8 (66.04 to 67.31 cm)
Item Description Button Front
Length (English) 38.25 in.
Length (Metric) 97.15 cm
Construction Polyester, Combed Cotton
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.