Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

REEP1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB187970 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
200 μg
50 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB187970 50 μg
NB301765 100 μL
NB187969 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. NB187970 Supplier Novus Biologicals Supplier No. NBP31464750UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

REEP1 Polyclonal antibody specifically detects REEP1 in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)

Specifications

Antigen REEP1
Applications Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Immunoprecipitation 1 to 5 μL/mg of lysate, Immunohistochemistry-Paraffin 1:50-1:200
Formulation PBS, pH 7.0 with 0.03% Proclin 300
Gene Alias C2orf23, chromosome 2 open reading frame 23, receptor accessory protein 1, receptor expression-enhancing protein 1, SPG31FLJ13110
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human REEP1 fragment
Purification Method Antigen and protein A Affinity-purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Neurodegeneration, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 65055
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.