Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

REEP2 Antibody (S326D-29), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB259314700 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB259314700 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB259314700 Supplier Novus Biologicals Supplier No. NBP259314AF700

Mouse Monoclonal Antibody

REEP2 Monoclonal specifically detects REEP2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen REEP2
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone S326D-29
Conjugate Alexa Fluor 700
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias C5orf19receptor expression-enhancing protein 2, chromosome 5 open reading frame 19, receptor accessory protein 2, SGC32445receptor expression enhancing protein 2
Gene Symbols REEP2
Host Species Mouse
Immunogen Fusion protein amino acids 111-254 (Cytoplasmic C-terminus) of mouse REEP2.
Purification Method Protein G purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 51308
Test Specificity Detects 30kDa. Cross-reacts with REEP1.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.