Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

REEP4 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB290166 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB290166 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB290166 Supplier Novus Biologicals Supplier No. NBP230361PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

REEP4 Polyclonal antibody specifically detects REEP4 in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative)

Specifications

Antigen REEP4
Applications Immunoprecipitation
Classification Polyclonal
Conjugate PerCP
Dilution Immunoprecipitation, Western Blot (Negative)
Formulation PBS
Gene Alias C8orf20chromosome 8 open reading frame 20, FLJ22246, FLJ22277, PP432, receptor accessory protein 4, receptor expression enhancing protein 4, receptor expression-enhancing protein 4
Host Species Rabbit
Immunogen Maps to a region between residue 200 to 225 of human Receptor Accessory Protein 4 using the numbering given in entry NP_079508.2 (GeneID 80346).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 80346
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.