Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Reg1A Antibody (431202), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB303357 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB303357 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB303357 Supplier Novus Biologicals Supplier No. FAB4937V

Rat Monoclonal Antibody

Reg1A Monoclonal antibody specifically detects Reg1A in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF

Specifications

Antigen Reg1A
Applications Western Blot, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 431202
Conjugate Alexa Fluor 405
Dilution Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias ICRF, Islet cells regeneration factor, Islet of Langerhans regenerating protein, lithostathine-1-alpha, P19, pancreatic stone protein, secretory, Pancreatic thread protein, protein-X, PSP, PSPS, PSPS1, PTP, REG, REG-1-alpha, Regenerating islet-derived protein 1-alpha, Regenerating protein I alpha
Host Species Rat
Immunogen E. coli-derived recombinant human Reg1A, Gln23-Asn166, Accession # P05451
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 5967
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.