Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Reg1B Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB323444 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB323444 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB323444 Supplier Novus Biologicals Supplier No. NBP299850MFV450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reg1B Polyclonal antibody specifically detects Reg1B in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)

Specifications

Antigen Reg1B
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution ELISA, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias lithostathine-1-beta, Pancreatic stone protein 2, pancreatic threadprotein), PSPS2PSP-2, regenerating islet-derived 1 beta, Regenerating islet-derived protein 1-beta, Regenerating protein I beta, REGH, REGLREG-1-beta, secretory pancreatic stone protein 2
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human Reg1B (Accession#: NP_006498.1; Met 1-Asn 166)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5968
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.