Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Reg1B Rabbit anti-Human, Clone: 8, Novus Biologicals™
SDP

Catalog No. NB021340 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB021340 100 μg
NB021341 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB021340 Supplier Novus Biologicals Supplier No. NBP289937100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Reg1B Monoclonal specifically detects Reg1B in Human samples. It is validated for Sandwich ELISA Capture.

Specifications

Antigen Reg1B
Applications ELISA
Classification Monoclonal
Clone 8
Conjugate Unconjugated
Dilution Sandwich ELISA Capture 1:250-1:2000
Formulation 0.2 um filtered solution in PBS
Gene Alias lithostathine-1-beta, Pancreatic stone protein 2, pancreatic threadprotein), PSPS2PSP-2, regenerating islet-derived 1 beta, Regenerating islet-derived protein 1-beta, Regenerating protein I beta, REGH, REGLREG-1-beta, secretory pancreatic stone protein 2
Host Species Rabbit
Immunogen Recombinant Human Reg1B Protein
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5968
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.