Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Reg2 Antibody (RM0312-5D45), DyLight 405, Novus Biologicals™
SDP

Catalog No. NBP212429V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP212429V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP212429V Supplier Novus Biologicals Supplier No. NBP212429V
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Reg2 Monoclonal specifically detects Reg2 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen Reg2
Applications Western Blot
Classification Monoclonal
Clone RM0312-5D45
Conjugate DyLight 405
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias regenerating islet-derived 2, RegII
Gene Symbols REG1B
Host Species Rat
Immunogen Mouse recombinant Reg2
Purification Method Protein A or G purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 5968
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.