Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Reg3A Antibody (010), PE, Novus Biologicals™
SDP

Catalog No. NB333131 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB333131 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB333131 Supplier Novus Biologicals Supplier No. NBP290468PE
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Reg3A Monoclonal antibody specifically detects Reg3A in Mouse samples. It is validated for ELISA

Specifications

Antigen Reg3A
Applications ELISA
Classification Monoclonal
Clone 010
Conjugate PE
Dilution ELISA
Formulation PBS
Gene Alias hepatocarcinoma-intestine-pancreas, HIPpancreatitis-associated protein, Human proislet peptide, pancreatic beta cell growth factor, Pancreatitis-associated protein 1, PAP homologous protein, PAP1FLJ18565, PAPPAP-H, PBCGF, proliferation-inducing protein 34, proliferation-inducing protein 42, Reg III-alpha, REG3, REG-3-alpha, regenerating islet-derived 3 alpha, regenerating islet-derived protein 3-alpha, Regenerating islet-derived protein III-alpha, REG-III
Host Species Rabbit
Immunogen Rabbit immunized with purified, recombinant Mouse Reg3A (Accession#: NP_035389.1, Met1-Gln175)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5068
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.