Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Reg3A Antibody (019), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB109750 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB109750 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB109750 Supplier Novus Biologicals Supplier No. NBP289856AF647
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Reg3A Monoclonal specifically detects Reg3A in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Reg3A
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 19
Conjugate Alexa Fluor 647
Formulation 50 mM Sodium Borate
Gene Alias hepatocarcinoma-intestine-pancreas, HIPpancreatitis-associated protein, Human proislet peptide, pancreatic beta cell growth factor, Pancreatitis-associated protein 1, PAP homologous protein, PAP1FLJ18565, PAPPAP-H, PBCGF, proliferation-inducing protein 34, proliferation-inducing protein 42, Reg III-alpha, REG3, REG-3-alpha, regenerating islet-derived 3 alpha, regenerating islet-derived protein 3-alpha, Regenerating islet-derived protein III-alpha, REG-III
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Reg3A (Q06141-1; Met 1-Asp 175).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5068
Test Specificity No cross-reactivity in ELISA with: Human REG3G Mouse REG3A Human REG1B
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.