Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Reg4 Antibody (244), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB306106 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB306106 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB306106 Supplier Novus Biologicals Supplier No. NBP289832APC
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Reg4 Monoclonal antibody specifically detects Reg4 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen Reg4
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 244
Conjugate APC
Dilution Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias Gastrointestinal secretory protein, GISPREG-4, Reg IV, regenerating gene type IV, regenerating islet-derived family, member 4, regenerating islet-derived protein 4, Regenerating islet-derived protein IV, REG-IV, RELPREG-like protein
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Reg4 (Accession#: NP_001152824.1; Met 1-Pro 158).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 83998
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.