Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Regulatory protein GAL4 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB336822 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB336822 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB336822 Supplier Novus Biologicals Supplier No. NBP276808PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Regulatory protein GAL4 Polyclonal antibody specifically detects Regulatory protein GAL4 in Yeast samples. It is validated for Western Blot

Specifications

Antigen Regulatory protein GAL4
Applications Western Blot
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot
Formulation PBS
Gene Alias GAL4, GAL81, YPL248C
Host Species Rabbit
Immunogen Partial recombinant protein made to a N terminal portion of the yeast GAL4 protein (amino acids 1-147) [UniProt P04386]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Yeast
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.