Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RelA/NFkB p65 Antibody (112A1021), DyLight 405, Novus Biologicals™
SDP

Catalog No. NBP227416V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227416V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP227416V Supplier Novus Biologicals Supplier No. NBP227416V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RelA/NFkB p65 Monoclonal specifically detects RelA/NFkB p65 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen RelA/NFkB p65
Applications Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 112A1021
Conjugate DyLight 405
Dilution Western Blot, Flow Cytometry, Immunohistochemistry-Paraffin
Gene Accession No. Q969V5
Gene Alias MGC131774, nf kb p65, NFkB p65, NF-kB p65, NFKB3v-rel avian reticuloendotheliosis viral oncogene homolog A (nuclear factor ofkappa light polypeptide gene enhancer in B-cells 3 (p65)), Nuclear factor NF-kappa-B p65 subunit, Nuclear factor of kappa light polypeptide gene enhancer in B-cells 3transcription factor p65, p65, RelA, rela p65, v-rel reticuloendotheliosis viral oncogene homolog A (avian), v-rel reticuloendotheliosis viral oncogene homolog A, nuclear factor of kappalight polypeptide gene enhancer in B-cells 3, p65
Gene Symbols RELA
Host Species Mouse
Immunogen This NfkB p65 antibody was raised against a synthetic peptide corresponding to amino acids 526-539 of human NF-kB (p65) protein.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology, Immune System Diseases, Immunology, Innate Immunity, Neurodegeneration, Neuroscience, Signal Transduction, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 5970
Test Specificity Cross reacts with Human, Mouse and Rat. Predicted to react with Cow, Gorilla, New World Monkey and Pig.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.