Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RelA/NFkB p65 Antibody (465003), HRP, Novus Biologicals™
SDP

Catalog No. NB347465 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB347465 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB347465 Supplier Novus Biologicals Supplier No. FAB50781H
Add to Cart
Add to Cart

Rat Monoclonal Antibody

RelA/NFkB p65 Monoclonal specifically detects RelA/NFkB p65 in Human samples. It is validated for CyTOF, Flow Cytometry (Intracellular Staining).

Specifications

Antigen RelA/NFkB p65
Applications CyTOF, Flow Cytometry (Intracellular Staining)
Classification Monoclonal
Clone 465003
Conjugate HRP
Dilution Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias MGC131774, nf kb p65, NFkB p65, NF-kB p65, NFKB3v-rel avian reticuloendotheliosis viral oncogene homolog A (nuclear factor ofkappa light polypeptide gene enhancer in B-cells 3 (p65)), Nuclear factor NF-kappa-B p65 subunit, Nuclear factor of kappa light polypeptide gene enhancer in B-cells 3transcription factor p65, p65, RelA, rela p65, v-rel reticuloendotheliosis viral oncogene homolog A (avian), v-rel reticuloendotheliosis viral oncogene homolog A, nuclear factor of kappalight polypeptide gene enhancer in B-cells 3, p65
Host Species Rat
Immunogen E. coli-derived recombinant human RelA/NF kappa B p65, Met1-Thr254, Accession # NP_068810
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology, Immune System Diseases, Immunology, Innate Immunity, Neurodegeneration, Neuroscience, Phospho Specific, Signal Transduction, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 5970
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.