Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RelA/NFkB p65 Antibody, Novus Biologicals™
SDP

Catalog No. NB1002176 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002176 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1002176 Supplier Novus Biologicals Supplier No. NB1002176
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 28 publications

RelA/NFkB p65 Polyclonal specifically detects RelA/NFkB p65 in Human, Mouse, Rat samples. It is validated for Chemotaxis, ChIP Assay, ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen RelA/NFkB p65
Applications Chemotaxis, ChIP Assay, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:400-1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Chromatin Immunoprecipitation (ChIP)
Formulation PBS, 100 ug BSA. with 0.1% Sodium Azide
Gene Accession No. Q04206
Gene Alias MGC131774, nf kb p65, NFkB p65, NF-kB p65, NFKB3v-rel avian reticuloendotheliosis viral oncogene homolog A (nuclear factor ofkappa light polypeptide gene enhancer in B-cells 3 (p65)), Nuclear factor NF-kappa-B p65 subunit, Nuclear factor of kappa light polypeptide gene enhancer in B-cells 3transcription factor p65, p65, RelA, rela p65, v-rel reticuloendotheliosis viral oncogene homolog A (avian), v-rel reticuloendotheliosis viral oncogene homolog A, nuclear factor of kappalight polypeptide gene enhancer in B-cells 3, p65
Gene Symbols RELA
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the C-terminus of human NFkB p65.
Molecular Weight of Antigen 69 kDa
Purification Method IgG purified
Quantity 1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology, Neurodegeneration, Neuroscience, Signal Transduction, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 5970
Test Specificity Reacts with NFkB p65
Target Species Human, Mouse, Rat
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.