Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Relaxin-3 Antibody (MM0526-7G14), DyLight 405, Novus Biologicals™
SDP

Catalog No. NBP211866V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP211866V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP211866V Supplier Novus Biologicals Supplier No. NBP211866V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Relaxin-3 Monoclonal specifically detects Relaxin-3 in Human samples. It is validated for Western Blot.

Specifications

Antigen Relaxin-3
Applications Western Blot
Classification Monoclonal
Clone MM0526-7G14
Conjugate DyLight 405
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias H3, insl7, relaxin 3, RXN3, ZINS4
Gene Symbols RLN3
Host Species Mouse
Immunogen Human recombinant Relaxin-3
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 117579
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.