Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Relaxin-3 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB343376 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB343376 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB343376 Supplier Novus Biologicals Supplier No. NBP241317PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Relaxin-3 Polyclonal antibody specifically detects Relaxin-3 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen Relaxin-3
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias H3, insl7, relaxin 3, RXN3, ZINS4
Host Species Rabbit
Immunogen Antibody was raised against a 19 amino acid peptide near the center of human RLN3. The immunogen is located within amino acids 80 - 130 of RLN3. Amino Acid Squence: LALTKSPQAFYRGRPSWQG
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 117579
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.