Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Relaxin R1/LGR7 Antibody (933344), PE/Atto594, Novus Biologicals™
SDP

Catalog No. NB310646 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB310646 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB310646 Supplier Novus Biologicals Supplier No. FAB8898PEATT594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Relaxin R1/LGR7 Monoclonal specifically detects Relaxin R1/LGR7 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen Relaxin R1/LGR7
Applications Flow Cytometry
Classification Monoclonal
Clone 933344
Conjugate PE/ATTO 594
Dilution Flow Cytometry
Formulation PBS
Gene Alias leucine-rich repeat-containing G protein-coupled receptor 7, Leucine-rich repeat-containing G-protein coupled receptor 7, LGR7.1, LGR7.10, LGR7LGR7.2, MGC138347, MGC142177, Relaxin family peptide receptor 1, relaxin receptor 1, relaxin/insulin-like family peptide receptor 1, RXFPR1
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human Relaxin R1, Met1-Ser398, Accession # Q9HBX9
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 59350
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.