Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Relaxin R1/LGR7 Antibody, Novus Biologicals™
SDP

Catalog No. NBP223674 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP223674 0.1 mL
NBP223674SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP223674 Supplier Novus Biologicals Supplier No. NBP223674
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

Relaxin R1/LGR7 Polyclonal specifically detects Relaxin R1/LGR7 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen Relaxin R1/LGR7
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot reported in scientific literature (PMID 31218471), Immunohistochemistry, Immunohistochemistry-Paraffin 1:100, Immunohistochemistry-Frozen reported in scientific literature (PMID 31218471)
Formulation Crude antibody whole antisera with 50% Glycerol with 0.025% Sodium Azide
Gene Accession No. NP_067647.2
Gene Alias leucine-rich repeat-containing G protein-coupled receptor 7, Leucine-rich repeat-containing G-protein coupled receptor 7, LGR7.1, LGR7.10, LGR7LGR7.2, MGC138347, MGC142177, Relaxin family peptide receptor 1, relaxin receptor 1, relaxin/insulin-like family peptide receptor 1, RXFPR1
Gene Symbols RXFP1
Host Species Rabbit
Immunogen Within the range of amino acids 694-732 of human RXFP1 protein were used as the immunogen for this antibody.
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 59350
Test Specificity The amino acid sequence used as immunogen for this RXFP1 antibody is 100% homologous in Human, Chimp and Orangutan, and 83% homologous in Horse, Bovine, Dog and Cat.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.