Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Relaxin R2/LGR8 Antibody (1013903) [Janelia Fluor™ 549], Novus Biologicals™
SDP

Catalog No. NB402159 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB402159 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB402159 Supplier Novus Biologicals Supplier No. FAB102501I
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Relaxin R2/LGR8 Monoclonal antibody specifically detects Relaxin R2/LGR8 in Human samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen Relaxin R2/LGR8
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 1013903
Conjugate Janelia Fluor 549
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias G protein coupled receptor affecting testicular descent, GPR106G-protein coupled receptor 106, GREATRelaxin family peptide receptor 2, INSL3R, leucine-rich repeat-containing G protein-coupled receptor 8, Leucine-rich repeat-containing G-protein coupled receptor 8, LGR8.1, LGR8G-protein coupled receptor affecting testicular descent, relaxin receptor 2, relaxin/insulin-like family peptide receptor 2, RXFPR2
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human Relaxin R2, Met1-Asn416, Accession # Q8WXD0
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 122042
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.