Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RelB Antibody, Novus Biologicals™
SDP

Catalog No. NBP122980 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP122980 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP122980 Supplier Novus Biologicals Supplier No. NBP122980
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RelB Polyclonal specifically detects RelB in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen RelB
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunoprecipitation 5-15 ug/mg lysate
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. NP_006500.2
Gene Alias IREL, I-REL, nuclear factor of kappalight polypeptide gene enhancer in B-cells 3, v-rel reticuloendotheliosis viral oncogene homolog B
Gene Symbols RELB
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 529 and 579 of human v-rel reticuloendotheliosis viral oncogene homolog B using the numbering given in entry NP_006500.2 (GeneID 5971).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5971
Test Specificity Human, Mouse. Based on 100% sequence identity, this antibody is predicted to react with Bovine, Dog, Horse, Rabbit, Guinea pig_10141, Panda, Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, White-tufted-ear marmoset, Crab-eating macaque, European domestic ferret, African elephant and Northern white-cheeked gibbon.
Target Species Human, Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.