Learn More
Promega ReliaPrep RNA Cell Miniprep System, 50 preps
Catalog No. prz6011 Shop All Promega Corporation Products
Change view
Click to view available options
:
ReliaPrep RNA Cell Miniprep System, 50 preps
Catalog No.
PRZ6011 ReliaPrep RNA Cell Miniprep System, 50 preps
1 options
Catalog No. PRZ6011 Supplier Promega Supplier No. Z6011
Only null left
Add to Cart
Add to Cart
The ReliaPrep RNA Miniprep Systems provide a fast and simple technique for preparation of intact total RNA from cultured cells or tissue in as little as 30 minutes from very small amounts of input material.
  • Efficiently Purify RNA from as Few as 100 Cells or 0.25mg Tissue
  • Simple, 30−minute protocol
  • Elutes in small volume, generating highly concentrated RNA
  • No phenol/chloroform or ß−mercaptoethanol required
Specifications
Content And Storage 15°C to 30°C
Quantity 50 Preps