Learn More
Description
Specifications
Specifications
| Antigen | RELL2 |
| Applications | Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, pH7.3. |
| Gene Alias | C5orf16, chromosome 5 open reading frame 16, FLJ90583, receptor expressed in lymphoid tissues like 2, RELT-like 2, RELT-like protein 2 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 44-303 of human RELL2 (NP_776189.3). |
| Purification Method | Affinity purified |
| Quantity | 100 μL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
