Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RELM beta Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB018129 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018129 0.025 mg
NB200204 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018129 Supplier Novus Biologicals Supplier No. NB200204SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RELM beta Polyclonal specifically detects RELM beta in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen RELM beta
Applications Immunoprecipitation, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500 -1:1000, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Formulation PBS with 0.02% Sodium Azide
Gene Alias C/EBP-epsilon regulated myeloid-specific secreted cysteine-rich proteinprecursor 2, CCRG, Colon and small intestine-specific cysteine-rich protein, Colon carcinoma-related gene protein, cysteine-rich secreted A12-alpha-like protein 1, Cysteine-rich secreted protein A12-alpha-like 1, Cysteine-rich secreted protein FIZZ2, FIZZ1, FIZZ2RELM-beta, found in inflammatory zone 1, HXCP2XCP2, RELMb, RELMbeta, resistin like beta, resistin-like beta, RETNL2
Gene Symbols RETNLB
Host Species Rabbit
Immunogen Reacts with the residues 32-46 of the mouse RELM beta protein
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 84666
Test Specificity Reacts with the residues 32-46 of the mouse RELM beta protein.
Target Species Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.