Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RELM beta Antibody (MRB 46L), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB172222 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB172222 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB172222 Supplier Novus Biologicals Supplier No. NBP280033C
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

RELM beta Monoclonal antibody specifically detects RELM beta in Mouse samples. It is validated for Western Blot, ELISA

Specifications

Antigen RELM beta
Applications Western Blot, ELISA
Classification Monoclonal
Clone MRB 46L
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias C/EBP-epsilon regulated myeloid-specific secreted cysteine-rich proteinprecursor 2, CCRG, Colon and small intestine-specific cysteine-rich protein, Colon carcinoma-related gene protein, cysteine-rich secreted A12-alpha-like protein 1, Cysteine-rich secreted protein A12-alpha-like 1, Cysteine-rich secreted protein FIZZ2, FIZZ1, FIZZ2RELM-beta, found in inflammatory zone 1, HXCP2XCP2, RELMb, RELMbeta, resistin like beta, resistin-like beta, RETNL2
Host Species Rat
Immunogen Recombinant mouseRELM beta.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 84666
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.