Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RELT/TNFRSF19L Antibody (238104), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB175151 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB175151 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB175151 Supplier Novus Biologicals Supplier No. FAB1385E
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

RELT/TNFRSF19L Monoclonal antibody specifically detects RELT/TNFRSF19L in Human samples. It is validated for Flow Cytometry

Specifications

Antigen RELT/TNFRSF19L
Applications Flow Cytometry
Classification Monoclonal
Clone 238104
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias Receptor expressed in lymphoid tissues, RELT tumor necrosis factor receptor, TNFRSF19LFLJ14993, tumor necrosis factor receptor superfamily member 19L, tumor necrosis factor receptor superfamily, member 19-like
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human RELT/TNFRSF19L, Met1-Ala160 (Arg127Gly, Arg129Gly), Accession # Q969Z4
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 84957
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.