Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RELT/TNFRSF19L Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP25685125UL
Description
RELT/TNFRSF19L Polyclonal specifically detects RELT/TNFRSF19L in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
RELT/TNFRSF19L | |
Polyclonal | |
Western Blot 0.4 μg/mL, Immunocytochemistry/Immunofluorescence 1-4 μg/mL | |
Receptor expressed in lymphoid tissues, RELT tumor necrosis factor receptor, TNFRSF19LFLJ14993, tumor necrosis factor receptor superfamily member 19L, tumor necrosis factor receptor superfamily, member 19-like | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
IgG |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
RELT | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:GGGSGINPAYRTEDANEDTIGVLVRLITEKKENAAALEELLKEYHSKQLVQTSHRPVSKLPPAPPNVPHIC | |
25 μL | |
Cancer | |
84957 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction