Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RELT/TNFRSF19L Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | RELT/TNFRSF19L |
---|---|
Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
RELT/TNFRSF19L Polyclonal specifically detects RELT/TNFRSF19L in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
RELT/TNFRSF19L | |
Polyclonal | |
Rabbit | |
Cancer | |
Receptor expressed in lymphoid tissues, RELT tumor necrosis factor receptor, TNFRSF19LFLJ14993, tumor necrosis factor receptor superfamily member 19L, tumor necrosis factor receptor superfamily, member 19-like | |
RELT | |
IgG | |
Affinity Purified |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
84957 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:GGGSGINPAYRTEDANEDTIGVLVRLITEKKENAAALEELLKEYHSKQLVQTSHRPVSKLPPAPPNVPHIC | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title