Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

REM2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NBP267107 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP267107 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP267107 Supplier Novus Biologicals Supplier No. NBP267107

Rabbit Polyclonal Antibody

REM2 Polyclonal specifically detects REM2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen REM2
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:20, Immunohistochemistry-Paraffin 1:200-1:400
Formulation PBS (pH7.4), 0.2% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias CD300LE, FLJ38964, GTP-binding protein REM 2, ICLM2, Rad and Gem-like GTP-binding protein 2, Rad and Gem-related 2 (rat homolog), RAS (RAD and GEM) like GTP binding 2, RAS (RAD and GEM)-like GTP binding 2
Gene Symbols REM2
Host Species Rabbit
Immunogen Synthetic peptide within Human REM2 aa 291-340/ 340. (SwissProt: Q8IYK8 Human)
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 161253
Target Species Human, Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.