Learn More
Thermo Scientific™ Columbia CNA Agar Base (Dehydrated)
Catalog No. R452962 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
Catalog No. Quantity
R452962 500 g
1 options
This item is not returnable. View return policy
Catalog No. R452962 Supplier Thermo Scientific™ Supplier No. R452962
Add to Cart
Add to Cart
This item is not returnable. View return policy
Selectively isolate gram positive cocci using Columbia CNA Agar Base.
Selectively isolate gram-positive cocci using Thermo Scientific™ Remel™ Columbia CNA Agar Base (Dehydrated). Columbia Agar Base was first described by Ellner, Stoessel, Drakeford, and Vasi in 1966 as an improved agar base1 which was capable of growing fastidious and nonfastidious organisms from clinical specimens. Ellner et al. incorporated colistin and nalidixic acid in the base to produce a selective medium for isolating gram-positive organisms (e.g., streptococci, staphylococci) from specimens containing mixed flora.
Carry out primary selective isolation of gram-positive cocci using Columbia CNA Agar Base.
  • Selective: Nalidixic acid and colistin inhibit most gram-negative bacilli.
  • Ease of Identification: Use of sheep blood enables the demonstration of hemolytic activity.
This medium contains peptones which supply growth factors such as nitrogen, carbon, vitamins, and trace elements essential for bacterial growth. Beef extract and corn starch serve as sources of energy and yeast extract supplies B-complex vitamins. Colistin and nalidixic acid inhibit most gram-negative bacilli. Sheep blood provides the X factor (hemin) necessary for the growth of many bacteria and enables demonstration of hemolytic activity.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Specifications
Description Columbia CNA Agar Base
Form Powder
Product Type Agar
Quantity 500 g
For Laboratory Use Only