Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Remel™ GC Agar Base
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
Form:
Powder
Description
Remel GC Agar Base is used with hemoglobin and GCHI enrichment for the isolation and cultivation of Neisseria/gonorrhoeae and other fastidious microorganisms.
Also available: GCHI Enrichment with Rehydrating Solution, Part No. R450411; VCN Selective Supplement, Part No. R450901; and VCNT Selective Supplement, Part No. R450921.
Specifications
Specifications
| Description | GC Agar Base |
| Form | Powder |
| Product Type | Agar |
| Quantity | 500 g |
For Laboratory Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction