Learn More
Thermo Scientific™ Middlebrook 7H9 Broth (Dehydrated)
Isolate and cultivate Mycobacterium spp. using Middlebrook 7H9 Broth.
$398.00 - $1832.00
Specifications
Description | Middlebrook 7H9 Broth |
---|---|
Form | Powder |
Product Type | Dehydrated Culture Media |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
R454012
|
Thermo Scientific™
R454012 |
500 g |
Each for $398.00
|
|
|||||
R454014
|
Thermo Scientific™
R454014 |
2.5 kg |
Each for $1,832.00
|
|
|||||
Description
Isolate and cultivate Mycobacterium spp. using Thermo Scientific™ Remel™ Middlebrook 7H9 Broth (Dehydrated). In 1958, Middlebrook and Cohn were the first to formulate this medium for the cultivation of mycobacteria1. This medium is found useful in the transfer of cultures to be tested with biochemical media2.
Isolate and cultivate Mycobacterium spp. using Middlebrook 7H9 Broth.
- Ease of Handling: Ease of transfer of cultures to be tested with biochemical media.
- Allows luxuriant growth of Mycobacterium
This medium is used for qualitative procedures. This medium contains inorganic salts which are essential for the growth of mycobacteria. Ammonium sulphate acts as a source of nitrogen. Biotin plays an important role in carboxylation and decarboxylation reactions, while pyridoxin is a precursor of enzymatic activity. The addition of ADC Enrichment provides a carbon source of energy, and catalase minimizes toxic effects.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Middlebrook 7H9 Broth | |
Powder | |
Dehydrated Culture Media |
For Laboratory Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.