Learn More
Thermo Scientific™ Nutrient Agar (Dehydrated)
Catalog No. r454182
Click to view available options
Quantity:
500 g
This item is not returnable. View return policy
Catalog No. R454182
Supplier: Thermo Scientific™ R454182
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Cultivate and maintain wide variety of microorganisms with Nutrient Agar.
Cultivate and maintain wide variety of microorganisms with Thermo Scientific™ Remel™ Nutrient Agar (Dehydrated). This is a non-selective medium which was formulated by APHA in 19171 as a standaridized medium for the use in the examination of water, wastewater, dairy products and various foods2-4. This medium is also used for the recovery of non-fastidious microorganisms and to cultivate and maintain stock cultures.
Cultivate and maintain wide variety of microorganisms with Nutrient Agar.
  • Nutrient Agar is a general-purpose medium used to subculture organisms for maintenance purposes or to check the purity of subcultures from isolation plates prior to biochemical or serological tests.
  • Nutrient Agar is suitable for teaching and demonstration purposes.
This medium is used for qualitative procedures. This medium contains beef extract that provides carbohydrates, vitamins, nitrogen compounds, and salts. Gelatin peptone provides nutrients in the form of amino acids and peptides. They are found to be nutritious as meat infusion.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Industrial Reference: AOAC, BAM, COMPF, SMD.
Description Nutrient Agar
Form Powder
Product Type Agar
Quantity 500 g
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.