Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Tryptic Soy Agar w/Lecithin, Polysorbate 80 (Dehydrated)

Catalog No. R455032
Click to view available options
Quantity:
500 g
2.5 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Detect and enumerate microorganisms present on surfaces of sanitary importance with this solid medium.

Detect and enumerate microorganisms present on surfaces of sanitary importance with Thermo Scientific™ Remel™ Tryptic Soy Agar w/Lecithin, Polysorbate 80 (Dehydrated).

Formulated by Weber and Black that lecithin added to an agar medium in sufficient concentration neutralizes quaternary ammonium compounds1. Brummer also tested the medium2. United States Pharmacopeia (USP)3 mentions Tryptin Soy Agar (TSA) that is prepared according to the formula for Soybean-Casein Digest Agar. Tryptic Soy Agar w/Lecithin, Polysorbate 80 (Dehydrated) is used in environmental air sampling procedures.

Detect microorganisms using the solid medium Tryptic Soy Agar w/Lecithin, Polysorbate 80.

  • Neutralization easiness: attained by lecithin and polysorbate 80 that are added to residual compounds present on the materials that are sampled.
  • Conforms to the formulation mentioned in the United States Pharmacopeia (USP)

Tryptic Soy Agar w/Lecithin, Polysorbate 80 contains casein peptone and soy peptone, sodium chloride, and agar that help in growth of bacteria, maintains osmotic equilibrium, and acts as a solidifying agent, respectively.

 

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description Tryptic Soy Agar w/Lecithin, Polysorbate 80
Form Powder
Product Type Agar
Quantity 500 g

For Laboratory Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.