Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Renalase Antibody (RNLS/1940), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB386897 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB386897 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB386897 Supplier Novus Biologicals Supplier No. NBP308450AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Renalase Monoclonal antibody specifically detects Renalase in Human samples. It is validated for ELISA

Specifications

Antigen Renalase
Applications ELISA
Classification Monoclonal
Clone RNLS/1940
Conjugate Alexa Fluor 532
Formulation 50mM Sodium Borate
Gene Alias C10orf59chromosome 10 open reading frame 59, EC 1.4, FLJ11218, MAO-C, Monoamine oxidase-C, RENALASE, renalase, FAD-dependent amine oxidase
Host Species Mouse
Immunogen Recombinant human Renalase protein fragment (around aa 34-235) (exact sequence is proprietary) (Uniprot: Q5VYX0)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 55328
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.