Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RENALASE Mouse anti-Human, Clone: 3A12F5, Proteintech
SDP

Catalog No. 501736255 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6255 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6255 Supplier Proteintech Group Inc Supplier No. 601281IG150UL
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody is specific to RNLS.

Renalase is a flavin adenine dinucleotide-dependent amine oxidase that is secreted into the blood from the kidney.

Specifications

Antigen RENALASE
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 3A12F5
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene RNLS
Gene Accession No. Q5VYX0
Gene Alias C10orf59, MAO C, Monoamine oxidase C, RENALASE, RNLS
Gene Symbols RNLS
Host Species Mouse
Immunogen RENALASE Fusion Protein Ag13061
Purification Method Protein G
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55328
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.