Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RENALASE Mouse anti-Human, Clone: 3A12F5, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
| Antigen | RENALASE |
|---|---|
| Clone | 3A12F5 |
| Concentration | 0.6 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6255
![]()
|
Proteintech Group Inc
601281IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This antibody is specific to RNLS.
Renalase is a flavin adenine dinucleotide-dependent amine oxidase that is secreted into the blood from the kidney.Specifications
| RENALASE | |
| 0.6 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| C10orf59, MAO C, Monoamine oxidase C, RENALASE, RNLS | |
| RNLS | |
| IgG1 | |
| Protein G | |
| Antibody |
| 3A12F5 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q5VYX0 | |
| 55328 | |
| RENALASE Fusion Protein Ag13061 | |
| Primary | |
| -20°C | |
| RNLS |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title