Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Renin Antibody (1), PerCP, Novus Biologicals™
SDP

Catalog No. NB332949 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB332949 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB332949 Supplier Novus Biologicals Supplier No. NBP290510PCP
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Renin Monoclonal antibody specifically detects Renin in Mouse samples. It is validated for ELISA, Immunohistochemistry (Paraffin)

Specifications

Antigen Renin
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1
Conjugate PerCP
Dilution ELISA, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias angiotensin-forming enzyme, Angiotensinogenase, EC 3.4.23, EC 3.4.23.15, FLJ10761, HNFJ2, Prorenin, renin, renin precursor, renal
Host Species Rabbit
Immunogen Rabbit immunized with purified, recombinant Mouse Renin (Accession#: NP_112469.1, Met 1-Arg 402)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology, metabolism
Primary or Secondary Primary
Gene ID (Entrez) 5972
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.