Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Renin R Antibody, DyLight 405, Novus Biologicals™
SDP

Catalog No. NB058138 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB058138 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB058138 Supplier Novus Biologicals Supplier No. NBP299111V
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Renin R Polyclonal specifically detects Renin R in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Renin R
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 405
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias APT6M8-9, ATP6M8-9MRXE, ATPase H(+)-transporting lysosomal accessory protein 2, ATPase H(+)-transporting lysosomal-interacting protein 2, ATPase, H+ transporting, lysosomal accessory protein 2, ATPase, H+ transporting, lysosomal interacting protein 2, CAPER, ELDF10, Embryonic liver differentiation factor 10, ER-localized type I transmembrane adaptor, H+ transporting, lysosomal (vacuolar proton pump) membrane sectorassociated protein M8-9, M8-9, MGC99577, MSTP009, N14F, renin receptor, vacuolar proton ATP synthase membrane sector associated protein M8-9, V-ATPase M8.9 subunit, XMRE
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human Renin R.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10159
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.