Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Renin R Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 7 publications

Supplier:  Novus Biologicals NBP190820

 View more versions of this product

Catalog No. NBP190820


Only null left
Add to Cart

Description

Description

Renin R Polyclonal specifically detects Renin R in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.
Specifications

Specifications

Renin R
Polyclonal
Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:20 - 1:50, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:20 - 1:50, Knockdown Validated
APT6M8-9, ATP6M8-9MRXE, ATPase H(+)-transporting lysosomal accessory protein 2, ATPase H(+)-transporting lysosomal-interacting protein 2, ATPase, H+ transporting, lysosomal accessory protein 2, ATPase, H+ transporting, lysosomal interacting protein 2, CAPER, ELDF10, Embryonic liver differentiation factor 10, ER-localized type I transmembrane adaptor, H+ transporting, lysosomal (vacuolar proton pump) membrane sectorassociated protein M8-9, M8-9, MGC99577, MSTP009, N14F, renin receptor, vacuolar proton ATP synthase membrane sector associated protein M8-9, V-ATPase M8.9 subunit, XMRE
Rabbit
Affinity Purified
RUO
10159
Human, Mouse, Rat
IgG
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown
Unconjugated
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
ATP6AP2
This antibody was developed against Recombinant Protein corresponding to amino acids:NSLSRNNEVDLLFLSELQVLHDISSLLSRHKHLAKDHSPDLYSLELAGLDEIGKRYGEDSEQFRDASKILVDALQKFADDMYSLYGGNAVVELVTVKSFDTSLIRKTRTILEAKQAKNPASPYNLAYKYNFEYSVVFN
0.1 mL
Primary
Specificity of human Renin R antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only